missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzenesulfinate, 98%, Thermo Scientific Chemicals

Catalog Number 11935761
Click to view available options
Quantity:
5 mL
25 mL
This item is not returnable. View return policy
670-98-4
C7H8O2S
156.2
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
2759784
methyl benzenesulfinate
COS(=O)C1=CC=CC=C1
This item is not returnable. View return policy
670-98-4
C7H8O2S
156.2
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
2759784
methyl benzenesulfinate
COS(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1940g/mL
Boiling Point 110.0°C to 112.0°C (16.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 mL
Specific Gravity 1.194
Formula Weight 156.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl benzenesulfinate
Show More Show Less
Product Identifier
  • Methyl benzenesulfinate, 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'