Learn More
Methyle6-(trifluoromethyl)nicotinate, 97%, ACROS Organics™
Brand: Acros Organics 437950050
Code : 29
Additional Details : CAS Number : 221313-10-6 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
Methyle6-(trifluoromethyl)nicotinate | |
≥96.0% | |
C8H6F3NO2 | |
MFCD03001346 |
221313-10-6 | |
97% | |
205.13 |
Specifications
Methyle6-(trifluoromethyl)nicotinate | |
58° to 62°C | |
221313-10-6 | |
Glass bottle | |
≥96.0% | |
C8H6F3NO2 | |
MFCD03001346 |
Off-white | |
5g | |
Authentic | |
Powder | |
97% | |
205.13 |