missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-methylnicotinate, 97%, Thermo Scientific Chemicals

Catalog Number p-4228812
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5470-70-2
C8H9NO2
151.16
MFCD00006340
VYPPZXZHYDSBSJ-UHFFFAOYSA-N
methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
231548
methyl 6-methylpyridine-3-carboxylate
CC1=NC=C(C=C1)C(=O)OC
This item is not returnable. View return policy
5470-70-2
C8H9NO2
151.16
MFCD00006340
VYPPZXZHYDSBSJ-UHFFFAOYSA-N
methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
231548
methyl 6-methylpyridine-3-carboxylate
CC1=NC=C(C=C1)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 31.0°C to 37.0°C
Color Colorless to Yellow
Boiling Point 108.0°C to 110.0°C (22.5 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2 g
Formula Weight 151.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 6-methylnicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 6-methylnicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'