missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle5-bromonicotinate, 97%, Maybridge™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 216.03 |
Percent Purity | 97% |
Chemical Name or Material | Methyle5-bromonicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'