missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-bromo-4-methyl-2-thiophenecarboxylate, 97%, Thermo Scientific™

Catalog Number p-2705413
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
54796-47-3
C7H7BrO2S
235.095
MFCD04038959
RJWZFHRMQRWEIH-UHFFFAOYSA-N
methyl 5-bromo-4-methyl-2-thiophenecarboxylate, methyl 5-bromo-4-methyl-2-thiophene carboxylate, acmc-209ljm, 5-bromo-2-methoxycarbonyl-4-methylthiophene, methyl 5-bromo-4-methyl-thiophene-2-carboxylate, 5-bromo-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 5-bromo-4-methyl-, methyl ester, 5-bromo-4-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 5-bromo-4-methyl-, methyl ester
2795489
methyl 5-bromo-4-methylthiophene-2-carboxylate
CC1=C(SC(=C1)C(=O)OC)Br
This item is not returnable. View return policy
54796-47-3
C7H7BrO2S
235.095
MFCD04038959
RJWZFHRMQRWEIH-UHFFFAOYSA-N
methyl 5-bromo-4-methyl-2-thiophenecarboxylate, methyl 5-bromo-4-methyl-2-thiophene carboxylate, acmc-209ljm, 5-bromo-2-methoxycarbonyl-4-methylthiophene, methyl 5-bromo-4-methyl-thiophene-2-carboxylate, 5-bromo-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 5-bromo-4-methyl-, methyl ester, 5-bromo-4-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 5-bromo-4-methyl-, methyl ester
2795489
methyl 5-bromo-4-methylthiophene-2-carboxylate
CC1=C(SC(=C1)C(=O)OC)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.1
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-4-methyl-2-thiophenecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'