missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle4-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | methyl 4-trifluoromethyl nicotinate, methyl 4-trifluoromethyl pyridine-3-carboxylate, methyl4-trifluoromethyl nicotinate, 4-trifluoromethyl-nicotinic acid methyl ester, pubchem23562, methyl 4-trifluoromethylnicotinate, methyl 4-trifluormethylpyridine-3-carboxylate, methyl 4-trifluoromethylpyridin-3-ylcarboxylate |
Formula Weight | 205.13 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 4-(trifluoromethyl)nicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'