missing translation for 'onlineSavingsMsg'
Learn More

Methyle4-(trifluoromethoxy)benzoate, 97%, Maybridge

Brand:  Maybridge TL00423EB

Code : 31

Additional Details : CAS Number : 780-31-4 Weight : 0.07500kg

Quantity 25g

 View more versions of this product

Product Code. 10492494

  • £55.00 / 25g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyle4-(trifluoromethoxy)benzoate
97.0
97%
CF3OC6H4C(O)OCH3
MFCD00041006
780-31-4
97%
C9H7F3O3
220.15

Specifications

Methyle4-(trifluoromethoxy)benzoate
780-31-4
97%
C9H7F3O3
220.15
25g
97.0
97%
CF3OC6H4C(O)OCH3
MFCD00041006
Videos
SDS