missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-imidazolecarboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-4089842
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17325-26-7
C5H6N2O2
126.12
MFCD00216584
DVLGIQNHKLWSRU-UHFFFAOYSA-N
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
565661
methyl 1H-imidazole-5-carboxylate
COC(=O)C1=CN=CN1
This item is not returnable. View return policy
17325-26-7
C5H6N2O2
126.12
MFCD00216584
DVLGIQNHKLWSRU-UHFFFAOYSA-N
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
565661
methyl 1H-imidazole-5-carboxylate
COC(=O)C1=CN=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154°C to 156°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 126.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-imidazolecarboxylate
compliance-icons
Product Identifier
  • Methyl 4-imidazolecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'