missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-cyanobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-4259016
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1129-35-7
C9H7NO2
161.16
MFCD00001823
KKZMIDYKRKGJHG-UHFFFAOYSA-N
4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i
70791
methyl 4-cyanobenzoate
COC(=O)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
1129-35-7
C9H7NO2
161.16
MFCD00001823
KKZMIDYKRKGJHG-UHFFFAOYSA-N
4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i
70791
methyl 4-cyanobenzoate
COC(=O)C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 67.0°C to 68.0°C
Color White
Boiling Point 142.0°C to 144.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula NCC6H4CO2CH3
Quantity 5 g
Beilstein 09,846
Formula Weight 161.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-cyanobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'