missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-3736906
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
32807-28-6
C5H7ClO3
150.56
MFCD00000938
HFLMYYLFSNEOOT-UHFFFAOYSA-N
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
36240
methyl 4-chloro-3-oxobutanoate
COC(=O)CC(=O)CCl
This item is not returnable. View return policy
32807-28-6
C5H7ClO3
150.56
MFCD00000938
HFLMYYLFSNEOOT-UHFFFAOYSA-N
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
36240
methyl 4-chloro-3-oxobutanoate
COC(=O)CC(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14.0°C to 16.0°C
Color Orange-Yellow to Yellow
Density 1.2800g/mL
Boiling Point 212.0°C to 213.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C(O)CH2COOCH3
Refractive Index 1.4560 to 1.4580
Quantity 25 mL
Specific Gravity 1.28
Solubility Information Solubility in water: 71g/L (20 c)
Formula Weight 150.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 4-chloroacetoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'