missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-iodobenzoate, 95%, ACROS Organics™

Brand:  Acros Organics 397070250

Code : 29

Additional Details : CAS Number : 19718-49-1 Weight : 0.07500kg

Packaging Glass bottle

 View more versions of this product

Product Code. 10396454

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Methyl 4-amino-3-iodobenzoate
95%
NH2(I)C6H3COOCH3
MFCD00090423
95%
C8H8INO2
277.06

Specifications

Methyl 4-amino-3-iodobenzoate
86.0° to 91.0°C
19718-49-1
Glass bottle
95%
C8H8INO2
277.06
Beige
25g
Authentic
Crystalline Powder
95%
NH2(I)C6H3COOCH3
MFCD00090423
Videos
SDS