missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 99%, Thermo Scientific™

Catalog Number p-3779028
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
159660-85-2
C14H18N2O2
246.31
MFCD00216930
HLKWMBMESNOAMS-UHFFFAOYSA-N
methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester
563385
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N
This item is not returnable. View return policy
159660-85-2
C14H18N2O2
246.31
MFCD00216930
HLKWMBMESNOAMS-UHFFFAOYSA-N
methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester
563385
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N
Melting Point 87.0°C to 89.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 246.31
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'