missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trifluoromethyl)phenylacetate, 97%, Acros Organics™

Brand:  Acros Organics 365600250

Code : 29

Additional Details : CAS Number : 62451-84-7 Weight : 0.07500kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10656884

  • £206.20 / 25g


Estimated Shipment
18-07-2018
Add to basket


Specifications

Chemical Identifiers

Methyl 3-(trifluoromethyl)phenylacetate
97%
C10H9F3O2
MFCD04039762
62451-84-7
97%
218.17

Specifications

Methyl 3-(trifluoromethyl)phenylacetate
25g
Authentic
1.4500 to 1.4520
97%
C10H9F3O2
MFCD04039762
Clear colorless
62451-84-7
Glass bottle
Liquid
97%
218.17
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 3-(trifluoromethyl)phenylacetate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS