missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trifluoromethyl)phenylacetate, 97%, Thermo Scientific™

Catalog Number p-2691731
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
62451-84-7
C10H9F3O2
218.17
MFCD04039762
GMEBDKGPPKAPEM-UHFFFAOYSA-N
2734603
methyl 2-[3-(trifluoromethyl)phenyl]acetate
COC(=O)CC1=CC(=CC=C1)C(F)(F)F
This item is not returnable. View return policy
62451-84-7
C10H9F3O2
218.17
MFCD04039762
GMEBDKGPPKAPEM-UHFFFAOYSA-N
2734603
methyl 2-[3-(trifluoromethyl)phenyl]acetate
COC(=O)CC1=CC(=CC=C1)C(F)(F)F
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4500 to 1.4520
Quantity 5 g
Synonym methyl 2-3-trifluoromethyl phenyl acetate, methyl 3-trifluoromethyl phenylacetate, methyl 3-trifluoromethylphenyl acetate, methyl 3-trifluoromethyl phenyl acetate, methyltrifluoromethylphenylacetate, methyl m-trifluoromethylphenylacetate, methyl 3-trifluoromethylphenylacetate, methyl-3-trifluoromethylphenylacetate, methyl2-3-trifluoromethyl phenyl acetate
Formula Weight 218.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3-(trifluoromethyl)phenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-(trifluoromethyl)phenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'