missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle3-methyl-4-nitrobenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 195.17 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-methyl-4-nitrobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'