missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxyacrylate, 95%, Thermo Scientific Chemicals

Catalog Number 10495115
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
34846-90-7
C5H8O3
116.13
MFCD00128206
AUTCCPQKLPMHDN-ONEGZZNKSA-N
methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
5323651
methyl (E)-3-methoxyprop-2-enoate
COC=CC(=O)OC
This item is not returnable. View return policy
34846-90-7
C5H8O3
116.13
MFCD00128206
AUTCCPQKLPMHDN-ONEGZZNKSA-N
methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
5323651
methyl (E)-3-methoxyprop-2-enoate
COC=CC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3.0°C to 4.0°C
Color Colorless
Density 1.0700g/mL
Boiling Point 169.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H3COCH=CHCOOCH3
Refractive Index 1.4490 to 1.4510
Quantity 500 mL
Specific Gravity 1.07
Viscosity 1.68 mPa.s (20°C)
Formula Weight 116.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 3-methoxyacrylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-methoxyacrylate
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'