Learn More
Methyle3-iodo-4-methoxybenzoate, 98%, ACROS Organics™
Brand: Acros Organics 399090050
Code : 29
Additional Details : CAS Number : 35387-93-0 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
Methyle3-iodo-4-methoxybenzoate | |
98% | |
CH3OC6H3(I)COOCH3 | |
MFCD00052925 |
≥97.5% | |
C9H9IO3 | |
292.07 |
Specifications
Methyle3-iodo-4-methoxybenzoate | |
Authentic | |
Crystalline Powder | |
98% | |
CH3OC6H3(I)COOCH3 | |
MFCD00052925 | |
91° to 95°C |
35387-93-0 | |
Glass bottle | |
≥97.5% | |
C9H9IO3 | |
292.07 | |
Off-white | |
5g |