missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2705298
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
C6H6O3S
MFCD00055642
This item is not returnable. View return policy
C6H6O3S
MFCD00055642
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyle3-hydroxythiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'