missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle3-hydroxybutanoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
CAS Min % | 97.0 |
Density | 1.0500g/mL |
Assay Percent Range | 97% |
Linear Formula | CH3CH(OH)CH2CO2CH3 |
Quantity | 100 g |
Specific Rotation Condition | - 21.50 (20.00°C neat) |
Formula Weight | 118.13 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-hydroxybutanoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'