missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxybenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2705292
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
13134-76-4
C10H8O3S
208.23
MFCD00067732
JVUOTPTULVZUTL-KTKRTIGZSA-N
methyl 3-hydroxybenzo b thiophene-2-carboxylate, methyl 3-hydroxybenzo b thiophene-3-carboxylate, 2-hydroxy methoxy methylidene-1-benzothiophen-3-one, 2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one, benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester, benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester, maybridge4_000295, methyl 3-hydroxy-benzothiophene2-carboxylate
83165
CO\C(O)=C1/SC2=CC=CC=C2C1=O
This item is not returnable. View return policy
13134-76-4
C10H8O3S
208.23
MFCD00067732
JVUOTPTULVZUTL-KTKRTIGZSA-N
methyl 3-hydroxybenzo b thiophene-2-carboxylate, methyl 3-hydroxybenzo b thiophene-3-carboxylate, 2-hydroxy methoxy methylidene-1-benzothiophen-3-one, 2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one, benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester, benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester, maybridge4_000295, methyl 3-hydroxy-benzothiophene2-carboxylate
83165
CO\C(O)=C1/SC2=CC=CC=C2C1=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl 3-hydroxy-1-benzothiophene-2-carboxylate
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'