Learn More
Thermo Scientific Maybridge Methyl 3-chlorothiophene-2-carboxylate, 10g, 97%
Brand: Thermo Scientific Maybridge GK01295EA
Methyl 3-chlorothiophene-2-carboxylate, 10g, 97%
Code : 31
Additional Details : CAS Number : 88105-17-3 Weight : 0.06000kg