Learn More
Thermo Scientific Maybridge Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate, 1g, 97%
Brand: Thermo Scientific Maybridge GK01138DA
Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate, 1g, 97%
Code : 31
Additional Details : CAS Number : 175137-08-3 Weight : 0.05100kg