missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-pyrazinecarboxylate, 98%, Acros Organics™

Brand:  Acros Organics 269590100

Code : 29

Additional Details : CAS Number : 16298-03-6 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10445852

  • £57.80 / 10g


Estimated Shipment
19-03-2018
Add to basket


Specifications

Chemical Identifiers

Methyl 3-amino-2-pyrazinecarboxylate
16298-03-6
98%
153.14
98%
98%
C6H7N3O2
MFCD00010097

Specifications

Methyl 3-amino-2-pyrazinecarboxylate
166.0° to 172.0°C
98%
Authentic
Crystalline Powder
98%
153.14
25, IV, 4412
Yellow
10g
16298-03-6
Glass bottle
98%
C6H7N3O2
MFCD00010097
Videos
SDS