Learn More
Methyle3-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate, 97%, Maybridge
Brand: Maybridge KM06349EB
Code : 31
Additional Details : CAS Number : 175203-23-3 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate | |
97.0 | |
97% | |
323.19 |
175203-23-3 | |
97% | |
C14H15BrN2O2 | |
MFCD00068047 |
Specifications
Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate | |
97.0 | |
97% | |
323.19 | |
25g |
175203-23-3 | |
97% | |
C14H15BrN2O2 | |
MFCD00068047 |