missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-1,3-benzoxazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-4228811
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
136663-21-3
C10H9NO3
191.19
MFCD07774245
UFLDNRAXHGPDOD-UHFFFAOYSA-N
methyl 2-methylbenzo d oxazole-5-carboxylate, 5-benzoxazolecarboxylicacid, 2-methyl-, methyl ester, acmc-1c498, methyl2-methyl-1,3-benzoxazole-5-carboxylate, methyl 2-methylbenzoxazole-5-carboxylate, 5-methoxycarbonyl-2-methyl-1,3-benzoxazole, methyl 2-methyl-1?3-benzoxazole-5-carboxylate, 2-methylbenzoxazole-5-carboxylic acid methyl ester, 5-benzoxazolecarboxylicacid,2-methyl-,methyl ester, 5-benzoxazolecarboxylic acid, 2-methyl-, methyl ester
12157466
methyl 2-methyl-1,3-benzoxazole-5-carboxylate
CC1=NC2=C(O1)C=CC(=C2)C(=O)OC
This item is not returnable. View return policy
136663-21-3
C10H9NO3
191.19
MFCD07774245
UFLDNRAXHGPDOD-UHFFFAOYSA-N
methyl 2-methylbenzo d oxazole-5-carboxylate, 5-benzoxazolecarboxylicacid, 2-methyl-, methyl ester, acmc-1c498, methyl2-methyl-1,3-benzoxazole-5-carboxylate, methyl 2-methylbenzoxazole-5-carboxylate, 5-methoxycarbonyl-2-methyl-1,3-benzoxazole, methyl 2-methyl-1?3-benzoxazole-5-carboxylate, 2-methylbenzoxazole-5-carboxylic acid methyl ester, 5-benzoxazolecarboxylicacid,2-methyl-,methyl ester, 5-benzoxazolecarboxylic acid, 2-methyl-, methyl ester
12157466
methyl 2-methyl-1,3-benzoxazole-5-carboxylate
CC1=NC2=C(O1)C=CC(=C2)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 67°C to 68°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 191.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-methyl-1,3-benzoxazole-5-carboxylate
compliance-icons
Product Identifier
  • Methyl 2-methyl-1,3-benzoxazole-5-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'