Learn More
Thermo Scientific Maybridge Methyl 2-chloro-6-methylpyrimidine-4-carboxylate, 25g, 97%
Brand: Thermo Scientific Maybridge SPB06334EB
Methyl 2-chloro-6-methylpyrimidine-4-carboxylate, 25g, 97%
Code : 31
Additional Details : CAS Number : 89793-11-3 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III