missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-butynoate, 97%, Thermo Scientific Chemicals

Catalog Number 10542241
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
23326-27-4
C5H6O2
98.1
MFCD00009273
UJQCANQILFWSDJ-UHFFFAOYSA-N
methyl 2-butynoate, methyl tetrolate, 2-butynoic acid, methyl ester, 2-butynoic acid methyl ester, tetrolic acid methyl ester, but-2-ynoic acid methyl ester, methyl2-butynoate, methyl 2-butynoate ?, pubchem12783, acmc-1ceil
141019
methyl but-2-ynoate
CC#CC(=O)OC
This item is not returnable. View return policy
23326-27-4
C5H6O2
98.1
MFCD00009273
UJQCANQILFWSDJ-UHFFFAOYSA-N
methyl 2-butynoate, methyl tetrolate, 2-butynoic acid, methyl ester, 2-butynoic acid methyl ester, tetrolic acid methyl ester, but-2-ynoic acid methyl ester, methyl2-butynoate, methyl 2-butynoate ?, pubchem12783, acmc-1ceil
141019
methyl but-2-ynoate
CC#CC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 76°C to 77°C (80.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C∽CCO2CH3
Refractive Index 1.4360 to 1.438
Quantity 5 g
Beilstein 02, III, 1449
Specific Gravity 0.98
Formula Weight 98.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 2-butynoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 2-butynoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'