missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle2,6-dichloroisonicotinate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Spezifikation
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | methyl 2,6-dichloroisonicotinate, methyl2,6-dichloroisonicotinate, 2,6-chloro-4-pyridinecarboxylic acid methyl ester, 2,6-dichloroisonicotinic acid methyl ester, methyl-2,6-dichloroisonicotinate, 4-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, methyl-2,6-dichlorisonicotinat, acmc-209yt4, xskghsuhoyebtk-uhfffaoysa, methyl 2,6-dichloro-isonicotinate |
Formula Weight | 206.03 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2, 6-dichloroisonicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'