missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylated Spirit (IDA 99), 99% (v/v), Pure, (Industrial Methylated Spirit, 74 0.P.)
44.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Specifications
Melting Point | -114.1°C |
Color | Colorless |
Boiling Point | 78.2°C |
Packaging | Amber glass bottle |
Quantity | 1 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'