missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylated Spirit (IDA 94), 94% (v/v), Pure, (Industrial Methylated Spirit 64 0.P.), Fisher Chemical™
34.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
5 L
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
Metal Can
Plastic Coated Glass Bottle
Specifications
Melting Point | -114°C |
Color | Colorless |
Boiling Point | 78.2°C |
Packaging | Amber Glass Bottle |
Quantity | 1 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'