missing translation for 'onlineSavingsMsg'
Learn More

Methyl Vinyl Ketone, Stabilized ca 95%, Thermo Scientific Chemicals

Catalog Number 10552331
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5
C4H6O
70.09
MFCD00008777
FUSUHKVFWTUUBE-UHFFFAOYSA-N
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
6570
CHEBI:48058
but-3-en-2-one
CC(=O)C=C
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5
C4H6O
70.09
MFCD00008777
FUSUHKVFWTUUBE-UHFFFAOYSA-N
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
6570
CHEBI:48058
but-3-en-2-one
CC(=O)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Colorless
Density 0.8640g/mL
Boiling Point 80.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range ca. 95%
Packaging Glass bottle
Linear Formula H2C=CHCOCH3
Refractive Index 1.4100 to 1.4120
Quantity 5 mL
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
Specific Gravity 0.864
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Formula Weight 70.09
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material Methyl vinyl ketone
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl vinyl ketone, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'