missing translation for 'onlineSavingsMsg'
Learn More

Methyl tricosanoate, 99+%, Thermo Scientific Chemicals

Catalog Number 10461810
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2433-97-8
C24H48O2
368.65
MFCD00009349
VORKGRIRMPBCCZ-UHFFFAOYSA-N
tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
75519
methyl tricosanoate
CCCCCCCCCCCCCCCCCCCCCCC(=O)OC
2433-97-8
C24H48O2
368.65
MFCD00009349
VORKGRIRMPBCCZ-UHFFFAOYSA-N
tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
75519
methyl tricosanoate
CCCCCCCCCCCCCCCCCCCCCCC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 53°C
Flash Point >113°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)21CO2CH3
Quantity 5 g
Formula Weight 368.64
Percent Purity 99+%
Chemical Name or Material Methyl tricosanoate
compliance-icons
Product Identifier
  • Methyl tricosanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'