missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazole-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog Number p-7055022
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
55842-56-3
C5H5NO2S
143.16
MFCD18075580
YRMZUOPMHMPNPG-UHFFFAOYSA-N
7567660
methyl 1,3-thiazole-2-carboxylate
COC(=O)C1=NC=CS1
This item is not returnable. View return policy
55842-56-3
C5H5NO2S
143.16
MFCD18075580
YRMZUOPMHMPNPG-UHFFFAOYSA-N
7567660
methyl 1,3-thiazole-2-carboxylate
COC(=O)C1=NC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym methyl thiazole-2-carboxylate, methyl 2-thiazolecarboxylate, 2-thiazolecarboxylic acid methyl ester, methyl2-thiazolecarboxylate
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl thiazole-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'