Learn More
Thermo Scientific Maybridge Methyl tetrahydrothiopyran-4-carboxylate, 10g, 97%
Brand: Thermo Scientific Maybridge CC61922EA
Methyl tetrahydrothiopyran-4-carboxylate, 10g, 97%
Code : 31
Additional Details : CAS Number : 128094-82-6 Weight : 0.06000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III