missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl tert-Butyl Ether, ULTRA RESI-ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Specifications
Formula Weight | 88.15 |
Grade | ULTRA RESI-ANALYZED™ |
Quantity | 1 L |
Flash Point | −27.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.74kg |
Chemical Name or Material | Methyl tert-Butyl Ether |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P362-Take off contaminated clothing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'