missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D, Thermo Scientific Chemicals

Catalog Number 10791541
Click to view available options
Quantity:
7.5 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.1900g/mL
Deuterium (D) 99.95% min.
Flash Point 88°C
Specific Gravity 1.19
Viscosity 2.4 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Quantity 7.5 mL
CAS 2206-27-1
Melting Point 20.2°C
Boiling Point 190.0°C
Molecular Formula C2H6OS
Linear Formula (CD3)2SO
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Molecular Weight (g/mol) 84.17
PubChem CID 75151
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 100.0 atom % D
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'