missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
50.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Melting Point | 18.4°C |
Acidity | 0.0002 meq/g max. |
Density | 1.1000g/mL |
Boiling Point | 189.0°C |
Quantity | 1 L |
Flash Point | 87°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.4770 to 1.4800 |
Assay Percent Range | 99.5% min. (GC) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'