missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-168662
44.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
EcoSafPak™ Plastic Bottle
Plastic Bottle
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 18.4°C
CAS Min % 99.9
Color Colorless
CAS Max % 100.0
Decomposition Information >190°C
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 500 mL
Flash Point 87°C
Packaging EcoSafPak™ Plastic Bottle
Physical Form Liquid
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.9+%
Residue after Evaporation 0.01% max.
Specific Gravity 1.1
Titratable Acid 0.001 meq/g max.
Linear Formula (CH3)2SO
Vapor Pressure Vapor Pressure: 0.55mbar at 20°C
Viscosity 1.987 mPa.s (25°C)
Beilstein 01, IV, 1277
Water 0.1% max. (Coulometric)
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS Reagent
Chemical Name or Material Methyl sulfoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'