Learn More
Thermo Scientific Acros Methyl sulfoxide 99.7% 1.1009g/mL m.p. 18.4°C b.p. 189°C (760) r.i. 1.477 Acros Organics
Brand:  Thermo Scientific Acros 127790500
Methyl sulfoxide 99.7% 1.1009g/mL m.p. 18.4°C b.p. 189°C (760) r.i. 1.477 Acros Organics
Code : 29
Additional Details : CAS Number : 67-68-5 Weight : 0.05500kg
Product Code. 10581561