missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl stearate, 99%, Thermo Scientific Chemicals
23.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 38°C to 42°C |
Boiling Point | 180°C to 182 (4 mm) |
Flash Point | >110°C (230°F) |
Assay Percent Range | 99% |
Quantity | 5 g |
Beilstein | 1786213 |
Solubility Information | Soluble in chloroform. Insoluble in water. |
Formula Weight | 298.51 |
Chemical Name or Material | Methyl Stearate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'