missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl (S)-4-Boc-morpholine-3-carboxylate, 97%

Brand:  Alfa Aesar™ H64218.03

Code : NEW

Additional Details : CAS Number : 215917-98-9 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15471319

  • £328.60 / 1g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Methyl (S)-4-Boc-morpholine-3-carboxylate
245.27
UN2811
(S)-4-Boc-morpholine-3-carboxylic acid methyl ester; 4-tert-Butyl 3-methyl (S)-morpholine-3,4-dicarboxylate
215917-98-9
MFCD09756519
15401005

Specifications

Methyl (S)-4-Boc-morpholine-3-carboxylate
215917-98-9
97%
MFCD09756519
15401005
(S)-4-Boc-morpholine-3-carboxylic acid methyl ester; 4-tert-Butyl 3-methyl (S)-morpholine-3,4-dicarboxylate
1g
Solid
245.27
UN2811
White
Videos
SDS