missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red sodium salt, ACS
Click to view available options
Quantity:
25 g
100 g
Description
Methyl Red sodium salt is used as a pH indicator. In microbiology, it is used to make agar plates in bacteriology. Further, it is used as a coloring agent and also finds use in mouthwash, toothpaste and in cosmetics. It is also employed as a rocket propellant. In addition to this, it is used in the preparation of reversible stain for rapid detection of stained cells in each specimen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beilstein | 4065488 |
Solubility Information | Soluble in water. Slightly soluble in ethanol. |
Formula Weight | 291.29 |
Grade | ACS |
Color | Brown to Red |
Quantity | 25 g |
Physical Form | Crystalline |
Chemical Name or Material | Methyl Red sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'