missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red, pure, indicator
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | 178.0°C to 182.0°C |
Linear Formula | (CH3)2NC6H4N=NC6H4CO2H |
Solubility Information | Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid |
Infrared Spectrum | Authentic |
Formula Weight | 269.3 |
Grade | Pure |
Packaging | Glass Bottle |
Specific Extinction | 650 min. (430nm, in buffer) |
Visual Transition Interval | from pH 4.2 (red) to pH 6.2 (yellow) |
Color | Red |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'