missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red, ACS
27.74 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
pH indicator in pH range 4.2 - 6.3Methyl red is used as a pH indicator. It is red in pH less than 4.4 and yellow in pH over 6.2. It is used to study its degradation using Fenton's reagent. In microbiology, it is used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 179°C to 182°C |
Beilstein | 750102 |
Solubility Information | Practically insoluble in water. Soluble in alcohol,acetic acidSoluble in ethanol. Insoluble in water. |
Formula Weight | 269.3 |
Grade | ACS |
Color | Maroon to Red |
Quantity | 25 g |
Physical Form | Crystalline |
Chemical Name or Material | Methyl Red, For analysis ACS |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'