missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

Catalog Number p-4090078
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
95715-86-9
C12H21NO5
259.30
MFCD00674041
ZNBUXTFASGDVCL-UHFFFAOYNA-N
r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate
688220
3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C
This item is not returnable. View return policy
95715-86-9
C12H21NO5
259.30
MFCD00674041
ZNBUXTFASGDVCL-UHFFFAOYNA-N
r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate
688220
3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0820g/mL
Boiling Point 89.0°C (0.8 mmHg)
Flash Point 93°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.082
Formula Weight 259.3
Percent Purity 95%
Chemical Name or Material Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate
compliance-icons
Product Identifier
  • Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'