missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-4380100
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6164-79-0
C6H6N2O2
138.13
TWIIRMSFZNYMQE-UHFFFAOYSA-N
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
72662
methyl pyrazine-2-carboxylate
COC(=O)C1=NC=CN=C1
This item is not returnable. View return policy
6164-79-0
C6H6N2O2
138.13
TWIIRMSFZNYMQE-UHFFFAOYSA-N
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
72662
methyl pyrazine-2-carboxylate
COC(=O)C1=NC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 59.5°C to 61.5°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 138.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl pyrazine-2-carboxylate
compliance-icons
Product Identifier
  • Methyl pyrazine-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'