missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl pentafluorophenyl carbonate, 97%, Thermo Scientific Chemicals
42.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Methyl pentafluorophenyl carbonate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl pentafluorophenyl carbonate
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Methyl pentafluorophenyl carbonate
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Melting Point | 27°C to 31°C |
Boiling Point | 84°C (3 mmHg) |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 2289710 |
Solubility Information | Insoluble in water. |
IUPAC Name | methyl (2,3,4,5,6-pentafluorophenyl) carbonate |
Formula Weight | 242.1 |
Percent Purity | 97% |
Chemical Name or Material | Methyl pentafluorophenyl carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'