missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Catalog Number 10702774
110.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
80-48-8
C8H10O3S
186.23
MFCD00008417
VUQUOGPMUUJORT-UHFFFAOYSA-N
methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester
6645
methyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
80-48-8
C8H10O3S
186.23
MFCD00008417
VUQUOGPMUUJORT-UHFFFAOYSA-N
methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester
6645
methyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.2% max.
Melting Point 24.0°C to 30.0°C
Color White to Yellow
Boiling Point 292.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO3CH3
Refractive Index 1.5162 to 1.5182
Quantity 2.5 kg
Beilstein 11, 99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene
Formula Weight 186.23
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl p-toluenesulfonate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl p-toluenesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'