missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%, ACROS Organics™

Brand:  Acros Organics 414960025

Code : 29

Additional Details : CAS Number : 99-75-2 Weight : 2.55000kg


Quantity 2.5kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10713624

  • £158.50 / 2.50kg


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

Methyl p-toluate
99-75-2
C9H10O2
150.18
99%
99%
CH3C6H4COOCH3
MFCD00008441

Specifications

Methyl p-toluate
220.0° to 223.0°C
2.5kg
99-75-2
Glass bottle
99%
C9H10O2
150.18
90°C
32.0° to 35.0°C
99%
1.0600g/mL
1.06
99%
CH3C6H4COOCH3
MFCD00008441
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl p-toluate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS