missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%, Thermo Scientific Chemicals

Catalog Number 10796953
Click to view available options
Quantity:
100 g
250 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
99-75-2
MFCD00008441
QSSJZLPUHJDYKF-UHFFFAOYSA-N
methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
7455
methyl 4-methylbenzoate
CC1=CC=C(C=C1)C(=O)OC
This item is not returnable. View return policy
99-75-2
MFCD00008441
QSSJZLPUHJDYKF-UHFFFAOYSA-N
methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
7455
methyl 4-methylbenzoate
CC1=CC=C(C=C1)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 32.0°C to 35.0°C
Density 1.0600g/mL
Boiling Point 220.0°C to 223.0°C
Flash Point 90°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Quantity 250 g
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol,ether
Molecular Weight (g/mol) 150.18
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl p-toluate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl p-toluate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'